Anti-CR1 Mouse Monoclonal Antibody [clone: Ber-MAC-DRC]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CR1
Clonality: Monoclonal
Clone: Ber-MAC-DRC
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB155-05
- DIABMOB155
- Antigen symbol:
- CR1
- CR1
- Antigen name:
- Complement Receptor Type 1
- Complement Receptor Type 1
- Antigen synonyms:
- CD35|C3BR|CR1|C3b/C4b receptor|KN|Complement receptor type 1|C4BR
- CD35|C3BR|CR1|C3b/C4b receptor|KN|Complement receptor type 1|C4BR
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- Ber-MAC-DRC
- Ber-MAC-DRC
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa