Anti-Cd34 Endothelial Cell Mouse Monoclonal Antibody [clone: QBEND/10]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cd34 Endothelial Cell
Clonality: Monoclonal
Clone: QBEND/10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB098
- DIABMOB098-05
- Antigen symbol:
- CD34
- Cd34 Endothelial Cell
- Antigen name:
- CD34 molecule
- Antigen synonyms:
- CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- QBEND/10
- QBEND/10
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa