Anti-IL2RA Mouse Monoclonal Antibody [clone: IL2R.1]
Type:
Antigen: IL2RA
Clonality: Monoclonal
Clone: IL2R.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
- Catalog No:
- DIABMOB254-05
- DIABMOB254
- Antigen symbol:
- IL2RA
- IL2RA
- Antigen name:
- Interleukin 2 Receptor Alpha
- Interleukin 2 Receptor Alpha
- Antigen synonyms:
- TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- IL2R.1
- IL2R.1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa