Anti-Cd20 B Cell Mouse Monoclonal Antibody [clone: L26]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cd20 B Cell
Clonality: Monoclonal
Clone: L26
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB004-05
- DIABMOB004
- Antigen symbol:
- Cd20 B Cell
- CD20
- Antigen name:
- CD20 molecule
- Antigen synonyms:
- Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- L26
- L26
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa