Anti-CD4 Mouse Monoclonal Antibody [clone: 1F6]
DIABPDM093
- Antigen name:CD4 molecule
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:1F6
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:L3T4|p55|CD4mut|Ly-4|W3/25
- Size:6 mL
- Pk:6 ml
Type:
Antigen: CD4
Clonality: Monoclonal
Clone: 1F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: