Anti-CD5 Rabbit Polyclonal Antibody
Type:
Antigen: CD5
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP099-05
- DIABRP099
- Antigen symbol:
- CD5
- CD5
- Antigen name:
- CD5 molecule
- CD5 molecule
- Antigen synonyms:
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit