Anti-CTSD Rabbit Polyclonal Antibody
Type:
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP004-05
- DIABRP004
- Antigen symbol:
- CTSD
- CTSD
- Antigen name:
- Cathepsin D
- Cathepsin D
- Antigen synonyms:
- lysosomal aspartyl protease|HEL-S-130P|ceroid-lipofuscinosis|lysosomal aspartyl peptidase|CPSD|CLN10|neuronal 10
- lysosomal aspartyl protease|HEL-S-130P|ceroid-lipofuscinosis|lysosomal aspartyl peptidase|CPSD|CLN10|neuronal 10
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit