Anti-CTNNA1 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CTNNA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP079-05
- DIABRP079
- Antigen symbol:
- CTNNA1
- CTNNA1
- Antigen name:
- Catenin Alpha 1
- Catenin Alpha 1
- Antigen synonyms:
- Alpha N-catenin|Alpha-catenin-related protein|CTNNA2|Catenin alpha2|Catenin alpha1|Renal carcinoma antigen NY-REN-13,Catenin-alpha1|alphaN-catenin|Catenin-alpha2|alphaE-catenin|Catenin alpha-1|CAPR|Cadherin-associated protein|Catenin alpha-2|Alpha E-catenin
- Alpha N-catenin|Alpha-catenin-related protein|CTNNA2|Catenin alpha2|Catenin alpha1|Renal carcinoma antigen NY-REN-13,Catenin-alpha1|alphaN-catenin|Catenin-alpha2|alphaE-catenin|Catenin alpha-1|CAPR|Cadherin-associated protein|Catenin alpha-2|Alpha E-catenin
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit