Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]
Type:
Antigen: CD5
Clonality: Monoclonal
Clone: 4C7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
- Catalog No:
- DIABMOB138
- DIABMOB138-05
- Antigen symbol:
- CD5
- CD5
- Antigen name:
- CD5 molecule
- CD5 molecule
- Antigen synonyms:
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 4C7
- 4C7
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa