Anti-CD10 Mouse Monoclonal Antibody [clone: 56C6]
Catalog # DIABPDM107
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Neprilysin
- Antigen symbol:MME
- Clonality:Monoclonal
- Clone:56C6
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:Calla|Common Acute Lymphoblastic Leukemia Antigen|Nep|Mme|Cd10|Membrane Metallo-Endopeptidase|Cluster Of Differentiation 10|Neutral Endopeptidase
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: CD10
Clonality: Monoclonal
Clone: 56C6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: