Anti-CD3 epsilon gamma delta/CD3 omega Mouse Monoclonal Antibody [clone: F7.2.38]
Type:
Antigen: CD3 epsilon gamma delta/CD3 omega
Clonality: Monoclonal
Clone: F7.2.38
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- DIABMOB422-05
- DIABMOB422
- Antigen symbol:
- CD3
- CD3 epsilon gamma delta/CD3 omega
- Antigen name:
- CD3 Molecule
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- F7.2.38
- F7.2.38
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1