Anti-CASP3 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP096
- DIABRP096-05
- Antigen symbol:
- CASP3
- CASP3
- Antigen name:
- Caspase 3
- Caspase 3
- Antigen synonyms:
- caspase-3|procaspase3|caspase 3|protein Yama|Yama|cysteine protease CPP32|PARP cleavage protease|CPP32|apoptosis-related cysteine protease|CPP32B|SCA-1|apopain|SREBP cleavage activity 1
- caspase-3|procaspase3|caspase 3|protein Yama|Yama|cysteine protease CPP32|PARP cleavage protease|CPP32|apoptosis-related cysteine protease|CPP32B|SCA-1|apopain|SREBP cleavage activity 1
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit