Anti-BCL6 Mouse Monoclonal Antibody () [clone: P1F6]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: BCL6
Clonality: Monoclonal
Clone: P1F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB131-05
- DIABMOB131
- Antigen symbol:
- BCL6
- BCL6
- Antigen name:
- B-cell lymphoma 6 protein
- B-cell lymphoma 6 protein
- Antigen synonyms:
- BCL5|LAZ3|BCL6A|ZNF51|ZBTB27
- BCL5|LAZ3|BCL6A|ZNF51|ZBTB27
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- P1F6
- P1F6
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1