Anti-CASP7 Mouse Monoclonal Antibody [clone: 7CSP01]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CASP7
Clonality: Monoclonal
Clone: 7CSP01
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB372-05
- DIABMOB372
- Antigen symbol:
- CASP7
- CASP7
- Antigen name:
- Caspase 7
- Caspase 7
- Antigen synonyms:
- Caspase 7 Apoptosis-related Cysteine Peptidase|CASP-7|MCH3|CASP7|ICE-LAP3|CMH-1
- Caspase 7 Apoptosis-related Cysteine Peptidase|CASP-7|MCH3|CASP7|ICE-LAP3|CMH-1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 7CSP01
- 7CSP01
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa