Anti-CALD1 Mouse Monoclonal Antibody [clone: h-CALD]
Type:
Antigen: CALD1
Clonality: Monoclonal
Clone: h-CALD
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
- Catalog No:
- DIABMOB316-05
- DIABMOB316
- Antigen symbol:
- CALD1
- CALD1
- Antigen name:
- Caldesmon 1
- Caldesmon 1
- Antigen synonyms:
- HCAD|caldesmon 1|NAG22|L-CAD|CDM|H-CAD|LCAD
- HCAD|caldesmon 1|NAG22|L-CAD|CDM|H-CAD|LCAD
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- h-CALD
- h-CALD
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa