Anti-CA15-3 Mouse Monoclonal Antibody [clone: DF3]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CA15-3
Clonality: Monoclonal
Clone: DF3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB134-05
- DIABMOB134
- Antigen symbol:
- Ca 15-3
- CA15-3
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DF3
- DF3
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa