Anti-BOB.1/OBF.1 Mouse Monoclonal Antibody () [clone: Wue-AC5]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: BOB.1/OBF.1
Clonality: Monoclonal
Clone: Wue-AC5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB445-05
- DIABMOB445
- Antigen symbol:
- Bob.1
- BOB.1/OBF.1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- Wue-AC5
- Wue-AC5
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a