Anti-BCL2 Mouse Monoclonal Antibody () [clone: 124]
DIABPDM016
- Antigen name:B-cell lymphoma 2
- Antigen symbol:BCL2
- Clonality:Monoclonal
- Clone:124
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:PPP1R50|Bcl-2
- Size:6 mL
- Pk:6 ml
Type:
Antigen: BCL2
Clonality: Monoclonal
Clone: 124
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0