Anti-Abeta Mouse Monoclonal Antibody [clone: BAM-10]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Abeta
Clonality: Monoclonal
Clone: BAM-10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB410
- DIABMOB410-05
- Antigen symbol:
- ABETA
- APP
- Antigen name:
- Amyloid beta
- Amyloid beta (A4) Precursor Protein
- Antigen synonyms:
- APP|CTFgamma|CVAP|ABETA|APPI|AD1|Peptidase nexin-II|amyloid beta (A4) precursor protein|Alzheimer disease amyloid A4 protein homolog|ABPP|Amyloid beta A4 protein|Amyloidogenic glycoprotein AG|AAA|PN2
- CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- BAM-10
- BAM-10
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1