Anti-ACTA1 Mouse Monoclonal Antibody [clone: HHF35]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: ACTA1
Clonality: Monoclonal
Clone: HHF35
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB002
- DIABMOB002-05
- Antigen symbol:
- ACTA1
- ACTA1
- Antigen name:
- Actin Alpha 1, Skeletal Muscle
- Actin Alpha 1, Skeletal Muscle
- Antigen synonyms:
- ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HHF35
- HHF35
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa