Anti-GFAP Rabbit Polyclonal Antibody
Catalog # DIABPDR028
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Glial fibrillary acidic protein
- Antigen symbol:GFAP
- Clonality:Polyclonal
- Host:Rabbit
- Antigen synonyms:FLJ45472|Intermediate filament protein.|Astrocyte|p-GFAP Ser38|GFAP phospho S38|GFAP|Glial Fibrillary Acidic Protein
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: