Anti-Fibroblast Mouse Monoclonal Antibody [clone: 5B5]
Catalog # DIABMOB197
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Fibroblast
- Clonality:Monoclonal
- Clone:5B5
- Host:Mouse
- Isotype:IgG1 kappa
- Size:1 mL
- Pk:1 ml
Specifications
About this item
Type:
Antigen: Fibroblast
Clonality: Monoclonal
Clone: 5B5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: