Anti-GLUT1 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: GLUT 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP128-05
- DIABRP128
- Antigen symbol:
- GLUT1
- GLUT1
- Antigen name:
- Glucose Transporter 1
- Glucose Transporter 1
- Antigen synonyms:
- EIG12|erythrocyte/brain|GLUT1DS|facilitated glucose transporter member 1|GLUT|glucose transporter type 1|GLUT-1|HTLVR|human T-cell leukemia virus (I and II) receptor|DYT18|GLUT1|PED|DYT9|hepG2 glucose transporter|solute carrier family 2|DYT17
- EIG12|erythrocyte/brain|GLUT1DS|facilitated glucose transporter member 1|GLUT|glucose transporter type 1|GLUT-1|HTLVR|human T-cell leukemia virus (I and II) receptor|DYT18|GLUT1|PED|DYT9|hepG2 glucose transporter|solute carrier family 2|DYT17
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit