Anti-Flt1/VEGFR1 Rabbit Polyclonal Antibody
Type:
Antigen: Flt1/VEGFR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP077
- DIABRP077-05
- Antigen symbol:
- Flt1/VEGFR1
- FLT1
- Antigen name:
- FMS-related tyrosine kinase 1
- Antigen synonyms:
- FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit