Anti-Granulocyte Mouse Monoclonal Antibody [clone: BM-2]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Granulocyte
Clonality: Monoclonal
Clone: BM-2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB268-05
- DIABMOB268
- Antigen name:
- Granulocyte
- Granulocyte
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- BM-2
- BM-2
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1