Anti-A1AT Rabbit Polyclonal Antibody
DIABPDR021
- Antibody type:Secondary
- Antigen name:Alpha-1-Antitrypsin
- Antigen symbol:A1AT
- Clonality:Polyclonal
- Host:Rabbit
- Antigen synonyms:alpha-1 protease inhibitor|alpha-1-antitrypsin|serine (or cysteine) proteinase inhibitor|SERPINA1|member 1|alpha-1-antiproteinase|clade A|serpin A1|protease inhibitor 1 (anti-elastase)
- Size:6 mL
- Pk:6 ml
Type:
Antigen: A1AT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: