Anti-THRB Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: THRB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP150
- DIABRP150-05
- Antigen symbol:
- THRB
- THRB
- Antigen name:
- Thyroid Hormone Receptor, beta
- Thyroid Hormone Receptor, beta
- Antigen synonyms:
- GRTH|ERBA-BETA|THRB2|NR1A2|ERBA2|MGC126109|thyroid hormone receptor|avian)|oncogene ERBA2|THRB1|beta|MGC126110|THR1|thyroid hormone receptor beta 1|avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|generalized resistance to thyroid hormone|beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|Thyroid hormone receptor
- GRTH|ERBA-BETA|THRB2|NR1A2|ERBA2|MGC126109|thyroid hormone receptor|avian)|oncogene ERBA2|THRB1|beta|MGC126110|THR1|thyroid hormone receptor beta 1|avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|generalized resistance to thyroid hormone|beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2|Thyroid hormone receptor
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit