Anti-Vascular Endothelial GF Receptor Mouse Monoclonal Antibody [clone: FLT-11]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Vascular Endothelial GF Receptor
Clonality: Monoclonal
Clone: FLT-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB457
- DIABMOB457-05
- Antigen name:
- Vascular Endothelial GF Receptor
- Vascular Endothelial GF Receptor
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- FLT-11
- FLT-11
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1