Anti-Vascular Endothelial GF Mouse Monoclonal Antibody [clone: VG1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Vascular Endothelial GF
Clonality: Monoclonal
Clone: VG1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB308-05
- DIABMOB308
- Antigen name:
- Vascular Endothelial GF
- Vascular Endothelial GF
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- VG1
- VG1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa