Anti-TOP2A Mouse Monoclonal Antibody [clone: SWT3D1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: TOP2A
Clonality: Monoclonal
Clone: SWT3D1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB243
- DIABMOB243-05
- Antigensymbol:
- TOP2A
- TOP2A
- Antikroppsnamn:
- Topoisomerase 2 alpha
- Topoisomerase 2 alpha
- Antigen synonymer:
- alpha isozyme|DNA topoisomerase 2-alpha|DNA gyrase|DNA topoisomerase (ATP-hydrolyzing)|TP2A|170 kD|DNA topoisomerase II|TOP2
- alpha isozyme|DNA topoisomerase 2-alpha|DNA gyrase|DNA topoisomerase (ATP-hydrolyzing)|TP2A|170 kD|DNA topoisomerase II|TOP2
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- SWT3D1
- SWT3D1
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa