Order Entry
Sweden
ContactUsLinkComponent
 
Anti-TNFA Mouse Monoclonal Antibody [clone: J1D9]
 
Anti-TNFA Mouse Monoclonal Antibody [clone: J1D9]

 

  • Catalog No:
  • DIABMOB314
  • DIABMOB314-05
  • Antigen symbol:
  • TNFA
  • TNFA
  • Antigen name:
  • Tumor Necrosis Factor Alpha
  • Tumor Necrosis Factor Alpha
  • Antigen synonyms:
  • tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
  • tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • J1D9
  • J1D9
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1