Anti-Thyroid Hormone Receptor Mouse Monoclonal Antibody [clone: 1718]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Thyroid Hormone Receptor
Clonality: Monoclonal
Clone: 1718
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Specifications
- Catalog No:
- DIABMOB350
- DIABMOB350-05
- Antigensymbol:
- THR
- THR
- Antikroppsnamn:
- Thyroid Hormone Receptor
- Thyroid Hormone Receptor
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- 1718
- 1718
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG2a
- IgG2a