Anti-CK10 Mouse Monoclonal Antibody [clone: DE-K10]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody recognizes keratin polypeptide of 56.5 kDa. It stains suprabasal cells in the epidermis, thymic Hassall’s bodies. Simple and glandular epithelia, transitional epithelium and adenocarcinomas are negative with this antibody.
Type:
Antigen: CK10
Clonality: Monoclonal
Clone: DE-K10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB055-05
- DIABMOB055
- Antigensymbol:
- CK10
- CK10
- Antikroppsnamn:
- Cytokeratin 10
- Cytokeratin 10
- Antigen synonymer:
- K10|Keratin-10|EHK|type I cytoskeletal 10|BIE|BCIE|CK-10
- K10|Keratin-10|EHK|type I cytoskeletal 10|BIE|BCIE|CK-10
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- DE-K10
- DE-K10
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa