Anti-Cytokeratin Wide Spectrum Rabbit Polyclonal Antibody
This antibody reacts with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa MW. This antibody is well suited for the staining of a broad spectrum of human keratins.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP010-05
- DIABRP010
- DIABPDR035
- Antigen symbol:
- CK Wide Spectrum
- CK Wide Spectrum
- CK Wide Spectrum
- Antigen name:
- Cytokeratin Wide Spectrum
- Cytokeratin Wide Spectrum
- Cytokeratin Wide Spectrum
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Rabbit