Anti-Cytokeratin (Basic) Mouse Monoclonal Antibody [clone: AE3]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody recognizes most of the basic (Type II) keratins. It recognizes keratin polypeptides of 58 and 65 to 67 kDa. In keratinized epidermis, 58 kDa keratin is present in the basal and suprabasal layers, while 65 to 67 kDa keratin is present in the cells above the basal layers. This antibody can be used to stain different cell layers in an epithelial tissue and various neoplasms of epithelial origin.
Type:
Antigen: Cytokeratin (Basic)
Clonality: Monoclonal
Clone: AE3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABPDM044
- DIABMOB093
- Antigen symbol:
- CK (Basic)
- CK (Basic)
- Antigen name:
- Type 2 CK
- Type 2 CK
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- AE3
- AE3
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa