Anti-CSTA Mouse Monoclonal Antibody [clone: WR-23/2/3/3]
This antibody reacts with 11-13 kDa protein, Cystatin A. Cystatins are endogenous inhibitors of lysosomal proteases that form a superfamily, which is classified into 1, 2, and 3 families. Cystatin A is thought to be a tumor suppressor, since an inverse correlation seems to exist between the level of Cystatin A and tumor progression. Cystatin A has been implicated in aging of tissues, Alzheimer disease, viral infection and inflammatory processes.
Type:
Antigen: CSTA
Clonality: Monoclonal
Clone: WR-23/2/3/3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
- Catalog No:
- DIABMOB409
- DIABMOB409-05
- Antigensymbol:
- CSTA
- CSTA
- Antikroppsnamn:
- Cystatin A
- Cystatin A
- Antigen synonymer:
- PSS4|Cystatin A|AREI|STF1|STFA
- PSS4|Cystatin A|AREI|STF1|STFA
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- WR-23/2/3/3
- WR-23/2/3/3
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1
- IgG1