Anti-ERCC1 Mouse Monoclonal Antibody [clone: 8F1]
This antibody reacts with a 33-36 kDa protein known as ERCC1 (excision repair cross complementing) polypeptide. ERCC1 is required for nucleotide excision repair of damaged DNA and is homologous to RAD10. In mammalian cells, XPG cleaves 3’ of the DNA lesion while ERCC1-XPF complex makes the 5’ incision.
Type:
Antigen: ERCC1
Clonality: Monoclonal
Clone: 8F1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
- Catalog No:
- DIABMOB336
- DIABMOB336-05
- DIABPDM151
- Antigen symbol:
- ERCC1
- ERCC1
- ERCC1
- Antigen name:
- Excision repair cross-complementation group 1
- Excision repair cross-complementation group 1
- Excision repair cross-complementation group 1
- Antigen synonyms:
- RAD10|COFS4|UV20
- RAD10|COFS4|UV20
- RAD10|COFS4|UV20
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 8F1
- 8F1
- 8F1
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b