Anti-CCND2 Mouse Monoclonal Antibody [clone: DCS-3.1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody is specific to human cyclin D2. Overexpression of cyclin D2 leads to acceleration of the G1 phase. The expression of cyclin D2 protein is more restricted than that of cyclin D1.
Type:
Antigen: CCND2
Clonality: Monoclonal
Clone: DCS-3.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB179
- DIABMOB179-05
- Antigen symbol:
- CCND2
- CCND2
- Antigen name:
- Cyclin D2
- Cyclin D2
- Antigen synonyms:
- G1/S-specific cyclin-D2|MPPH3|G1/S-specific cyclin D2|MGC102758|KIAK0002
- G1/S-specific cyclin-D2|MPPH3|G1/S-specific cyclin D2|MGC102758|KIAK0002
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DCS-3.1
- DCS-3.1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a