Anti-CCND1 Mouse Monoclonal Antibody [clone: DCS-6]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody is specific to human cyclin D1. The cyclin D1 gene is located on 11q13 and has been shown to be amplified in a variety of tumors. Maximum expression of cyclin D1 occurs at a critical point in mid to late G1 phase of the cell cycle.
Type:
Antigen: CCND1
Clonality: Monoclonal
Clone: DCS-6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB178-05
- DIABMOB178
- DIABPDM046
- Antigen symbol:
- CCND1
- CCND1
- CCND1
- Antigen name:
- Cyclin D1
- Cyclin D1
- Cyclin D1
- Antigen synonyms:
- BCL1|PRAD1 oncogene|PRAD1|BCL-1 oncogene|G1/S-specific cyclin-D1|B-cell lymphoma 1 protein|U21B31|B-cell CLL/lymphoma 1|D11S287E
- BCL1|PRAD1 oncogene|PRAD1|BCL-1 oncogene|G1/S-specific cyclin-D1|B-cell lymphoma 1 protein|U21B31|B-cell CLL/lymphoma 1|D11S287E
- BCL1|PRAD1 oncogene|PRAD1|BCL-1 oncogene|G1/S-specific cyclin-D1|B-cell lymphoma 1 protein|U21B31|B-cell CLL/lymphoma 1|D11S287E
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- DCS-6
- DCS-6
- DCS-6
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a