Anti-CCNB1 Mouse Monoclonal Antibody [clone: V152]
Catalog # DIABMOB264
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Cyclin B1
- Antigen symbol:CCNB1
- Clonality:Monoclonal
- Clone:V152
- Host:Mouse
- Antigen synonyms:CCNB
- Size:1 mL
- Pk:1 ml
Specifications
About this item
This antibody is specific to a protein of 62 kDa identified as cyclin B1. Cyclin B1 expression is restricted to a specific short period of the cell cycle with cyclin B1 expression detected earlier and peaking in concentration before cyclin B2 expression. In a majority of tumors, positive staining with cyclin B corresponds to the proliferative status of the tumor.
Type:
Antigen: CCNB1
Clonality: Monoclonal
Clone: V152
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: