Anti-Epithelial Antigen Mouse Monoclonal Antibody [clone: VU-1D9]
This antibody reacts with two glycoproteins of 34 and 49 kDa present on the epithelial cells. It stains with simple epithelia and basal layers of stratified non-keratinized squamous epithelium.
Type:
Antigen: Epithelial Antigen
Clonality: Monoclonal
Clone: VU-1D9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
- Catalog No:
- DIABMOB062
- DIABPDM077
- DIABMOB062-05
- Antigen name:
- Epithelial Antigen
- Epithelial Antigen
- Epithelial Antigen
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- VU-1D9
- VU-1D9
- VU-1D9
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa