Anti-EMA Mouse Monoclonal Antibody [clone: E29]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody reacts with an antigen of 265-400 kDa belonging to a heterogeneous group of heavily glycosylated proteins called human milk fat globule proteins. It stains both normal and neoplastic cells. Among normal epithelia, it reacts strongly with mammary epithelium and glandular epithelia but shows a patchy staining with squamous epithelium.
Type:
Antigen: EMA
Clonality: Monoclonal
Clone: E29
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB401
- DIABMOB401-05
- Antigen symbol:
- EMA
- EMA
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- E29
- E29
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa