Anti-EGFR Mouse Monoclonal Antibody [clone: 111.6]
This antibody is specific to a 170 kDa protein. EGF-Receptor is a member of the type I family of growth factor receptors. The antibody recognizes an epitope mapping in the extracellular domain of EGFR. Overexpression of EGFR has been reported in tumors of breast, brain, bladder, lung, stomach, head and neck, esophagus, cervix, vulva, ovary and endometrium. EGFR is over expressed in squamous cell carcinoma.
Type:
Antigen: EGFR
Clonality: Monoclonal
Clone: 111.6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- DIABMOB267-05
- DIABMOB267
- DIABPDM132
- Antigen symbol:
- EGFR
- EGFR
- EGFR
- Antigen name:
- Epidermal growth factor receptor
- Epidermal growth factor receptor
- Epidermal growth factor receptor
- Antigen synonyms:
- PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 111.6
- 111.6
- 111.6
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1