Anti-EBV Cocktail Mouse Monoclonal Antibody [clone: CS1-CS4]
This antibody is specific to 60 kDa latent membrane protein (LMP-1) encoded by the BNLF1 gene of the EBV. Each clone reacts with different epitopes on the hydrophilic C-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis.
Type:
Antigen: EBV Cocktail
Clonality: Monoclonal
Clone: CS1-CS4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- DIABPDM128
- DIABMOB194-05
- DIABMOB194
- Antigen name:
- EBV Cocktail
- EBV Cocktail
- EBV Cocktail
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- CS1-CS4
- CS1-CS4
- CS1-CS4
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1