Anti-CK5 Mouse Monoclonal Antibody [clone: XM26]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody reacts with cytokeratin 5 intermediate filament protein. Basal cells of human epidermis express acidic cytokeratin 14 and basic cytokeratin 5. Cytokeratin 5 is a 58 kDa protein, which is closely related to cytokeratin 6. Cytokeratin 5 and 6 are expressed in basal cell epithelioma, basal cells of prostate, urothelium, vagina, squamous cell carcinomas of skin, tongue, epiglottis and rectal-anal region. Cytokeratin 5 can be useful in the distinction of mesotheliomas from most adenocarcinomas.
Type:
Antigen: CK5
Clonality: Monoclonal
Clone: XM26
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABPDM139
- DIABMOB361
- DIABMOB361-05
- Antigen symbol:
- CK5
- CK5
- CK5
- Antigen name:
- Cytokeratin 5
- Cytokeratin 5
- Cytokeratin 5
- Antigen synonyms:
- CK-5|58 kda cytokeratin|cytokeratin-5|type-II keratin Kb5|Dowling-Meara/Kobner/Weber-Cockayne types)|type II cytoskeletal 5|epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types|keratin 5 (epidermolysis bullosa simplex|keratin
- CK-5|58 kda cytokeratin|cytokeratin-5|type-II keratin Kb5|Dowling-Meara/Kobner/Weber-Cockayne types)|type II cytoskeletal 5|epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types|keratin 5 (epidermolysis bullosa simplex|keratin
- CK-5|58 kda cytokeratin|cytokeratin-5|type-II keratin Kb5|Dowling-Meara/Kobner/Weber-Cockayne types)|type II cytoskeletal 5|epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types|keratin 5 (epidermolysis bullosa simplex|keratin
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- XM26
- XM26
- XM26
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
Specifications
Frequently Bought Together