Anti-SRCAP Rabbit Polyclonal Antibody
BOSSBS-12119R
: Bioss
- Antibody type:Primary
- Antigen name:Snf2-related CREBBP activator Protein
- Antigen symbol:SRCAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:FLJ44499|KIAA0309|helicase SRCAP|domino homolog 1|EAF1|SWR1|domino homolog 2|Swi2/Snf2-related ATPase homolog domino homolog 1|Snf2-related CREBBP activator protein|DOMO1|Snf2-related CBP activator protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: SRCAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat