Anti-CACNA2D1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11981R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, alpha 2/delta subunit 1
- Antigen symbol:CACNA2D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:CACNA2D1|Voltage gated calcium channel subunit alpha 2/delta 1|Voltage-dependent calcium channel subunit delta-1|MHS3|L type calcium channel subunit alpha 2|CACNL2A|MHS 3|CA2D1_HUMAN|Voltage dependent calcium channel subunit alpha 2/delta 1|CCHL2A|CACN A2|H_DJ0560O14.1|CACNA2|Calcium channel L type DHPR alpha 2 subunit|Calcium channel voltage dependent alpha 2/delta subunit 1|Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit|Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta|WUGSC:H_DJ0560O14.1.|Dihydropyridine receptor alpha 2 subunit|Calcium channel L type alpha 2 polypeptide|Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat