Order Entry
Sweden
ContactUsLinkComponent
Anti-CAPS1 Rabbit Polyclonal Antibody
Anti-CAPS1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11812R
Supplier:  Bioss
Anti-CAPS1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11812R
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Calcyphosine
  • Antigensymbol:
    CAPS1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Antigen synonymer:
    CAPS-1|CADPS1|KIAA1121|Ca2+-dependent secretion activator|calcium-dependent secretion activator 1|CAPS|Ca2+-regulated cytoskeletal protein|calcium-dependent activator protein for secretion 1|Ca2+-dependent activator protein for secretion
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Rening:
    Purified by Protein A
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CAPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat