Order Entry
Sweden
ContactUsLinkComponent
 
Anti-NEFL Mouse Monoclonal Antibody (CF568) [clone: NFL/736]
 :  Biotium
 
Anti-NEFL Mouse Monoclonal Antibody (CF568) [clone: NFL/736]
 :  Biotium

 

  • Catalog No:
  • BNC680736-500
  • BNC680736-100
  • Antigen symbol:
  • NEFL
  • NEFL
  • Antigen name:
  • Neurofilament
  • Neurofilament
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone NFL/736
  • Clone NFL/736
  • Reactivity:
  • Pig, Chicken, Human, Bovine, Rat
  • Pig, Chicken, Human, Bovine, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 521461
  • 521461
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide