Order Entry
Sweden
ContactUsLinkComponent
Anti-BRCA2 Rabbit Polyclonal Antibody
Anti-BRCA2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1210R
Supplier:  Bioss
Anti-BRCA2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1210R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Breast Cancer Susceptibility Protein 2
  • Antigen symbol:
    BRCA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    675
  • Antigen synonyms:
    BRCA1/BRCA2-containing complex|breast cancer type 2 susceptibility protein|subunit 2|early onset|fanconi anemia group D1 protein|breast cancer 2 tumor suppressor|breast cancer susceptibility protein BRCA2|breast and ovarian cancer susceptibility gene
  • Storage buffer:
    Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4 °C
  • Immunogen:
    1-50/3418
  • Purification:
    Purified by Protein A
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Involved in double-strand break repair and/or homologous recombination. Binds RAD51 and potentiates recombinational DNA repair by promoting assembly of RAD51 onto single-stranded DNA (ssDNA). Acts by targeting RAD51 to ssDNA over double-stranded DNA, enabling RAD51 to displace replication protein-A (RPA) from ssDNA and stabilizing RAD51-ssDNA filaments by blocking ATP hydrolysis. Part of a PALB2-scaffolded HR complex containing RAD51C and which is thought to play a role in DNA repair by HR. May participate in S phase checkpoint activation. Binds selectively to ssDNA, and to ssDNA in tailed duplexes and replication fork structures. May play a role in the extension step after strand invasion at replication-dependent DNA double-strand breaks; together with PALB2 is involved in both POLH localization at collapsed replication forks and DNA polymerization activity. In concert with NPM1, regulates centrosome duplication. Interacts with the TREX-2 complex (transcription and export complex 2) subunits PCID2 and DSS1, and is required to prevent R-loop-associated DNA damage and thus transcription-associated genomic instability. Silencing of BRCA2 promotes R-loop accumulation at actively transcribed genes in replicating and non-replicating cells, suggesting that BRCA2 mediates the control of R-loop associated genomic instability, independently of its known role in homologous recombination (PubMed:24896180).

Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: BRCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat